Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005738.1 | NP_001005738.1 | 2 | 660 | ||
NM_001462.3 | NP_001453.1 | 2 | 758 | |||
XM_006723120.3 | XP_006723183.1 | 5 | 1491 | |||
XM_017026542.1 | XP_016882031.1 | 4 | 1335 | |||
GenBank mRNA | AF054013.1 | - | 1 | 615 | ||
AK290557.1 | - | 3 | 806 | |||
BC029125.2 | - | 1 | 629 | 5180065 | ||
BC071722.1 | - | 2 | 930 | 5212947 | ||
D10922.1 | - | 2 | 649 | |||
M76672.1 | - | 1 | 615 | |||
M84562.1 | - | 2 | 1387 | |||
M88107.1 | - | 2 | 654 | |||
U81501.1 | - | 1 | 635 | |||
X63819.1 | - | 2 | 768 |
Set Membership: |
G-protein coupled receptor |