Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004958.3 | NP_004949.1 | 2 | 172 | ||
XM_005263438.2 | XP_005263495.1 | 2 | 179 | |||
XM_011541166.2 | XP_011539468.1 | 2 | 172 | |||
XM_017000901.1 | XP_016856390.1 | 1 | 32 | |||
XM_017000902.1 | XP_016856391.1 | 2 | 172 | |||
GenBank mRNA | AB209995.1 | - | 2 | 153 | ||
AK302863.1 | - | 2 | 164 | |||
AK304273.1 | - | 2 | 161 | |||
BC117166.1 | - | 1 | 69 | 40125717 | ||
BP250183.1 | - | 2 | 177 | |||
DC403129.1 | - | 2 | 163 | |||
L34075.1 | - | 2 | 130 | |||
U88966.1 | - | 2 | 130 |