Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318521.1 | NP_001305450.1 | 3 | 339 | ||
NM_001318523.1 | NP_001305452.1 | 1 | 176 | |||
NM_006733.3 | NP_006724.2 | 2 | 433 | |||
XM_005262111.2 | XP_005262168.1 | 3 | 303 | |||
XM_011530894.2 | XP_011529196.1 | 3 | 332 | |||
XM_011530895.2 | XP_011529197.1 | 4 | 619 | |||
XM_011530897.2 | XP_011529199.1 | 4 | 494 | |||
XM_011530899.2 | XP_011529201.1 | 2 | 300 | |||
XM_017029383.1 | XP_016884872.1 | 3 | 332 | |||
GenBank mRNA | AK302986.1 | - | 3 | 332 | ||
BC012462.2 | - | 1 | 177 | 4537558 | ||
DA736853.1 | - | 3 | 339 | |||
X97249.1 | - | 2 | 432 |