Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002033.3 | NP_002024.1 | 1 | 1507 | ||
GenBank mRNA | BC136373.1 | - | 1 | 1387 | 9020360 | |
BC136374.1 | - | 1 | 1387 | 9020361 | ||
M58596.1 | - | 1 | 906 | |||
M58597.1 | - | 1 | 1397 |
Gene Symbol: LOC105369438 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135093.1 | 5 | 1009 |