Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACKR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122951.2 | NP_001116423.1 | 1 | 917 | ||
NM_002036.3 | NP_002027.2 | 2 | 1683 | |||
GenBank mRNA | AF030521.1 | - | 1 | 742 | ||
AK291593.1 | - | 2 | 1683 | |||
AK309521.1 | - | 1 | 1186 | |||
AY167991.1 | - | 1 | 736 | |||
BC095496.1 | - | 1 | 739 | |||
BM671230.1 | - | 1 | 338 | |||
BX537430.1 | - | 3 | 1580 | |||
CA435450.1 | - | 1 | 338 | |||
HE572751.1 | - | 2 | 736 | |||
JX081310.1 | - | 1 | 742 | |||
U01839.1 | - | 1 | 917 |