Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FYB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243093.1 | NP_001230022.1 | 2 | 310 | ||
NM_001465.4 | NP_001456.3 | 2 | 357 | |||
NM_199335.3 | NP_955367.1 | 2 | 357 | |||
XM_006714464.2 | XP_006714527.1 | 2 | 342 | |||
XM_006714465.2 | XP_006714528.1 | 3 | 695 | |||
XM_006714466.2 | XP_006714529.1 | 2 | 471 | |||
XM_011514008.2 | XP_011512310.1 | 4 | 795 | |||
XM_011514009.1 | XP_011512311.1 | 4 | 716 | |||
XM_011514010.1 | XP_011512312.1 | 3 | 434 | |||
XM_011514011.2 | XP_011512313.1 | 2 | 318 | |||
XM_011514012.2 | XP_011512314.1 | 2 | 318 | |||
XM_011514013.2 | XP_011512315.1 | 2 | 319 | |||
XM_017009316.1 | XP_016864805.1 | 2 | 317 | |||
GenBank mRNA | AF001862.1 | - | 1 | 261 | ||
AF198052.1 | - | 1 | 194 | |||
AK130159.1 | - | 2 | 352 | |||
AK290403.1 | - | 2 | 352 | |||
AK297077.1 | - | 2 | 310 | |||
AV761195.1 | - | 2 | 341 | |||
BC015933.1 | - | 2 | 286 | |||
BC117449.1 | - | 2 | 256 | 40126000 | ||
BC143645.1 | - | 2 | 256 | 9052159 | ||
BP265190.1 | - | 2 | 357 | |||
BX647195.1 | - | 2 | 342 | |||
BX647196.1 | - | 2 | 342 | |||
CD691549.1 | - | 2 | 367 | |||
U93049.1 | - | 1 | 224 |