Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000810.3 | NP_000801.1 | 11 | 1637 | ||
NM_001165037.1 | NP_001158509.1 | 11 | 1503 | |||
XM_005268258.2 | XP_005268315.1 | 11 | 1515 | |||
XM_006720459.2 | XP_006720522.1 | 11 | 1419 | |||
XM_017022055.1 | XP_016877544.1 | 10 | 5650 | |||
GenBank mRNA | A28104.1 | - | 11 | 1402 | ||
AK124673.1 | - | 1 | 1190 | |||
AK126891.1 | - | 1 | 3190 | |||
AK290453.1 | - | 11 | 1300 | |||
AK294303.1 | - | 8 | 960 | |||
AK303742.1 | - | 6 | 808 | |||
BC111979.1 | - | 10 | 1249 | 8327447 | ||
BC113422.1 | - | 10 | 1249 | 8322474 | ||
BT009830.1 | - | 9 | 1105 | |||
L08485.1 | - | 11 | 1410 |