Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002043.4 | NP_002034.3 | 1, 2 | 240 | ||
GenBank mRNA | BC130352.1 | - | 1, 2 | 159 | 40146383 | |
BC130354.1 | - | 1, 2 | 159 | 40146385 | ||
BX490975.1 | - | 1, 2 | 248 | |||
M86868.1 | - | 1, 2 | 186 |