Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002052.4 | NP_002043.2 | 2, 3 | 1229 | ||
GenBank mRNA | AY740706.1 | - | 1, 2 | 621 | ||
BC101580.1 | - | 1, 2 | 698 | 8069086 | ||
BC105108.1 | - | 1, 2 | 696 | 8144111 | ||
BC143434.1 | - | 1, 2 | 698 | 9051942 | ||
D78260.1 | - | 1, 2 | 910 | |||
L34357.1 | - | 1, 2 | 858 |