Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001483.2 | NP_001474.1 | 3 | 292 | ||
GenBank mRNA | AF029786.1 | - | 3 | 267 | ||
AJ001259.1 | - | 3 | 259 | |||
AK315648.1 | - | 3 | 277 | |||
BC000732.2 | - | 3 | 265 | 2959662 | ||
BC001837.2 | - | 3 | 265 | 2959662 | ||
BC030821.1 | - | 3 | 274 | 4808962 | ||
BT007112.1 | - | 3 | 259 | |||
CR407612.1 | - | 3 | 259 | |||
CX166452.1 | - | 3 | 292 |