Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004120.4 | NP_004111.2 | 9 | 1722 | ||
GenBank mRNA | AK122866.1 | - | 5 | 3427 | ||
AK226018.1 | - | 9 | 1648 | |||
AK314325.1 | - | 9 | 1677 | |||
BC022272.1 | - | 9 | 1677 | 4688776 | ||
BC073163.1 | - | 9 | 1659 | 6151056 | ||
CR457062.1 | - | 8 | 1408 | |||
M55543.1 | - | 9 | 1564 |