Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001487.3 | NP_001478.2 | 4 | 429 | ||
NR_037655.1 | 3 | 356 | ||||
NR_037656.1 | 4 | 692 | ||||
NR_037657.1 | 3 | 619 | ||||
GenBank mRNA | AI673441.1 | - | 1 | 158 | ||
AI816035.1 | - | 1 | 150 | |||
AK307069.1 | - | 3 | 700 | |||
BC130640.1 | - | 4 | 394 | 40146671 | ||
BC132795.1 | - | 4 | 394 | 40146817 | ||
BQ899204.1 | - | 4 | 692 | |||
BU601406.1 | - | 4 | 439 | |||
CN263524.1 | - | 3 | 403 | |||
D64007.1 | - | 4 | 425 | |||
S82447.1 | - | 4 | 382 |