Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021078.2 | NP_066564.2 | 3 | 651 | ||
XM_006721817.3 | XP_006721880.1 | 3 | 680 | |||
XM_006721818.3 | XP_006721881.1 | 3 | 372 | |||
XM_017024448.1 | XP_016879937.1 | 3 | 623 | |||
GenBank mRNA | AF029777.1 | - | 3 | 615 | ||
BC032743.1 | - | 3 | 628 | 5575574 | ||
BC039907.1 | - | 3 | 643 | 6064838 | ||
BC105977.1 | - | 3 | 620 | 6137532 |