Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115156.1 | NP_001108628.1 | 10 | 1396 | ||
NM_001494.3 | NP_001485.2 | 11 | 1531 | |||
XM_017016071.1 | XP_016871560.1 | 11 | 1226 | |||
GenBank mRNA | AB451295.1 | - | 11 | 1240 | ||
AB451429.1 | - | 11 | 1240 | |||
AF144713.1 | - | 8 | 1000 | |||
AK094671.1 | - | 3 | 1648 | |||
AK122841.1 | - | 5 | 2704 | |||
AK297177.1 | - | 12 | 1498 | |||
AK297554.1 | - | 10 | 1281 | |||
AK309360.1 | - | 12 | 1447 | |||
AK312485.1 | - | 11 | 1416 | |||
BC005145.2 | - | 11 | 1324 | 3504736 | ||
BC024168.1 | - | 5 | 469 | |||
BT006868.1 | - | 11 | 1240 | |||
BX648038.1 | - | 10 | 1077 | |||
CR457073.1 | - | 11 | 1240 | |||
D13988.1 | - | 11 | 1260 | |||
EU794614.1 | - | 11 | 1240 | |||
Y13286.1 | - | 11 | 1392 |