Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000821.6 | NP_000812.2 | 11 | 1648 | ||
NM_001142269.3 | NP_001135741.1 | 10 | 1477 | |||
XM_005264259.4 | XP_005264316.1 | 11 | 1607 | |||
XM_011532764.2 | XP_011531066.1 | 10 | 1443 | |||
XM_011532765.2 | XP_011531067.1 | 9 | 1282 | |||
XM_017003803.1 | XP_016859292.1 | 10 | 1470 | |||
GenBank mRNA | AK130139.1 | - | 2 | 240 | ||
AK222827.1 | - | 11 | 1608 | |||
AK225663.1 | - | 11 | 1607 | |||
AK297397.1 | - | 10 | 1417 | |||
AK298982.1 | - | 10 | 1302 | |||
AK313126.1 | - | 11 | 1616 | |||
BC013979.2 | - | 11 | 1570 | 4129593 | ||
EU847509.1 | - | 11 | 1545 | |||
L17128.1 | - | 11 | 1697 | |||
M81592.1 | - | 11 | 1628 |