Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000821.6 | NP_000812.2 | 6 | 818 | ||
NM_001142269.3 | NP_001135741.1 | 5 | 647 | |||
XM_005264259.4 | XP_005264316.1 | 6 | 777 | |||
XM_017003803.1 | XP_016859292.1 | 5 | 640 | |||
GenBank mRNA | AK222827.1 | - | 6 | 778 | ||
AK225663.1 | - | 6 | 777 | |||
AK297397.1 | - | 5 | 587 | |||
AK298982.1 | - | 5 | 541 | |||
BC013979.2 | - | 6 | 740 | 4129593 | ||
EU847509.1 | - | 6 | 715 | |||
L17128.1 | - | 6 | 867 | |||
M81592.1 | - | 6 | 798 |