Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145666.1 | NP_001139138.1 | 7 | 1330 | ||
NM_001145667.1 | NP_001139139.1 | 8 | 1363 | |||
NM_012201.5 | NP_036333.2 | 8 | 1363 | |||
NR_027264.1 | 8 | 1363 | ||||
GenBank mRNA | AK025457.1 | - | 5 | 756 | ||
AK027032.1 | - | 8 | 1324 | |||
AK304156.1 | - | 7 | 1300 | |||
BC060822.1 | - | 8 | 1334 | 5296427 | ||
U28811.1 | - | 8 | 1336 | |||
U64791.1 | - | 8 | 1316 |