Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLP1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002062.4 | NP_002053.3 | 10 | 1029 | ||
NR_136562.1 | 10 | 1029 | ||||
NR_136563.1 | 10 | 1029 | ||||
XM_017010750.1 | XP_016866239.1 | 9 | 1564 | |||
XM_017010751.1 | XP_016866240.1 | 8 | 1506 | |||
GenBank mRNA | BC112126.1 | - | 10 | 1041 | 8327594 | |
BC113493.1 | - | 10 | 1029 | 8322545 | ||
KR138540.1 | - | 10 | 995 | |||
L23503.1 | - | 10 | 994 | |||
U01104.1 | - | 10 | 1011 | |||
U01156.1 | - | 10 | 969 | |||
U01157.1 | - | 10 | 969 | |||
U10037.1 | - | 10 | 1095 |
Set Membership: |
G-protein coupled receptor |