Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000824.4 | NP_000815.1 | 5 | 602 | ||
NM_001166060.1 | NP_001159532.1 | 5 | 585 | |||
NM_001166061.1 | NP_001159533.1 | 5 | 602 | |||
XM_017008034.1 | XP_016863523.1 | 4 | 396 | |||
XM_017008035.1 | XP_016863524.1 | 5 | 579 | |||
GenBank mRNA | AF094754.1 | - | 5 | 565 | ||
AF094755.1 | - | 5 | 565 | |||
AK290617.1 | - | 5 | 536 | |||
BC032635.1 | - | 5 | 541 | 5247588 | ||
CD013911.1 | - | 4 | 439 | |||
DA528659.1 | - | 5 | 602 | |||
U33267.1 | - | 5 | 476 |