Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000405.4 | NP_000396.2 | 3 | 574 | ||
GenBank mRNA | AK225978.1 | - | 3 | 502 | ||
AK312494.1 | - | 3 | 469 | |||
BC009273.2 | - | 3 | 438 | 4053681 | ||
BX473154.1 | - | 3 | 552 | |||
L01439.1 | - | 3 | 499 | |||
M76477.1 | - | 3 | 499 | |||
X16087.1 | - | 3 | 367 | |||
X61094.1 | - | 2 | 284 | |||
X61095.1 | - | 3 | 430 | |||
X62078.1 | - | 3 | 467 |