Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253846.1 | NP_001240775.1 | 3 | 138 | ||
NM_001500.3 | NP_001491.1 | 3 | 392 | |||
XM_006715066.2 | XP_006715129.1 | 3 | 348 | |||
XM_011514500.1 | XP_011512802.1 | 3 | 105 | |||
XM_011514502.2 | XP_011512804.1 | 3 | 348 | |||
XM_011514503.2 | XP_011512805.1 | 3 | 348 | |||
XM_011514505.1 | XP_011512807.1 | 3 | 348 | |||
XM_011514506.1 | XP_011512808.1 | 3 | 348 | |||
XM_011514507.1 | XP_011512809.1 | 3 | 348 | |||
GenBank mRNA | AF040260.1 | - | 3 | 138 | ||
AF042377.1 | - | 3 | 255 | |||
AK313953.1 | - | 3 | 332 | |||
BC000117.1 | - | 3 | 357 | 3351070 | ||
BQ923510.1 | - | 392 | ||||
CR541929.1 | - | 3 | 180 | |||
CR541947.1 | - | 3 | 180 |