Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006877.3 | NP_006868.3 | 7, 8 | 811 | ||
GenBank mRNA | BC008281.1 | - | 7, 8 | 811 | 3635871 |