Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297571.1 | NP_001284500.1 | 5 | 440 | ||
NM_002075.3 | NP_002066.1 | 5 | 440 | |||
XM_011520953.2 | XP_011519255.1 | 5 | 372 | |||
GenBank mRNA | AB209283.1 | - | 4 | 1581 | ||
AB593098.1 | - | 5 | 366 | |||
AB593099.1 | - | 5 | 365 | |||
AF501884.1 | - | 4 | 230 | |||
AK313091.1 | - | 5 | 320 | |||
AU143349.1 | - | 5 | 440 | |||
BC000115.1 | - | 5 | 353 | 3351520 | ||
BC002454.2 | - | 5 | 360 | 3342882 | ||
BC015920.1 | - | 5 | 319 | 4648399 | ||
BT009800.1 | - | 4 | 230 | |||
M31328.1 | - | 4 | 236 |