Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005275.3 | NP_005266.2 | 2, 3 | 1630 | ||
XM_005249015.3 | XP_005249072.1 | 3, 4 | 1093 | |||
GenBank mRNA | AK026391.1 | - | 2, 3 | 292 | ||
AK298042.1 | - | 2, 3 | 469 | |||
AK301738.1 | - | 2, 3 | 220 | |||
AK302480.1 | - | 1, 2 | 358 | |||
AK316391.1 | - | 2, 3 | 296 | |||
BC013959.2 | - | 2, 3 | 279 | 2961417 | ||
BC018366.1 | - | 2, 3 | 1210 | 3924808 | ||
BT006648.1 | - | 2, 3 | 239 | |||
L25665.1 | - | 2, 3 | 193 |