Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000825.3 | NP_000816.4 | 2, 3 | 1941 | ||
NM_001083111.1 | NP_001076580.1 | 3, 4 | 1072 | |||
GenBank mRNA | BC126437.1 | - | 2, 3 | 763 | 8992153 | |
BC126463.1 | - | 2, 3 | 762 | 8992179 | ||
CB243067.1 | - | 1, 2 | 222 | |||
M12578.1 | - | 3, 4 | 263 | |||
X01059.1 | - | 2, 3 | 1305 |