Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256486.1 | NP_001243415.1 | 13 | 4061 | ||
NM_001256487.1 | NP_001243416.1 | 13 | 4089 | |||
NM_001256488.1 | NP_001243417.1 | 12 | 3966 | |||
NM_004487.4 | NP_004478.3 | 13 | 4046 | |||
XM_005247371.4 | XP_005247428.1 | 13 | 4521 | |||
XM_005247372.4 | XP_005247429.1 | 13 | 4097 | |||
XM_005247373.1 | XP_005247430.1 | 12 | 3913 | |||
XM_006713587.1 | XP_006713650.1 | 13 | 4036 | |||
XM_006713588.1 | XP_006713651.1 | 13 | 4021 | |||
XM_006713589.1 | XP_006713652.1 | 13 | 4018 | |||
XM_006713590.1 | XP_006713653.1 | 12 | 3913 | |||
XM_006713591.1 | XP_006713654.1 | 11 | 3796 | |||
XM_011512699.2 | XP_011511001.1 | 13 | 3988 | |||
XM_017006189.1 | XP_016861678.1 | 13 | 4506 | |||
XM_017006190.1 | XP_016861679.1 | 13 | 4673 | |||
XM_017006191.1 | XP_016861680.1 | 12 | 4398 | |||
XM_017006192.1 | XP_016861681.1 | 13 | 4082 | |||
XM_017006193.1 | XP_016861682.1 | 13 | 4079 | |||
XM_017006194.1 | XP_016861683.1 | 12 | 3974 | |||
XM_017006195.1 | XP_016861684.1 | 7 | 3943 | |||
GenBank mRNA | AB371588.1 | - | 13 | 4034 | ||
AB593126.1 | - | 12 | 3940 | |||
BC065301.1 | - | 12 | 3837 | |||
D25542.1 | - | 13 | 4044 | |||
X75304.1 | - | 13 | 4034 |