Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007240.2 | NP_001007241.2 | 9 | 1574 | ||
NM_001007241.2 | NP_001007242.2 | 8 | 1133 | |||
NM_001007242.2 | NP_001007243.2 | 7 | 1124 | |||
NM_001502.3 | NP_001493.2 | 8 | 1565 | |||
XM_017023155.1 | XP_016878644.1 | 8 | 1318 | |||
GenBank mRNA | AB035541.1 | - | 7 | 1354 | ||
AB035542.1 | - | 6 | 913 | |||
AK293384.1 | - | 8 | 1321 | |||
AK312277.1 | - | 8 | 1452 | |||
BC032693.2 | - | 6 | 971 | 5229838 | ||
BX647831.1 | - | 9 | 1462 | |||
BX648023.1 | - | 8 | 1459 | |||
BX648201.1 | - | 7 | 1011 | |||
BX649067.1 | - | 8 | 1020 | |||
CR749600.1 | - | 8 | 1524 | |||
D38225.1 | - | 7 | 925 | |||
U36221.1 | - | 8 | 1434 |