Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006143.2 | NP_006134.1 | 4 | 386 | ||
XM_011520623.2 | XP_011518925.1 | 4 | 887 | |||
XM_011520624.2 | XP_011518926.1 | 4 | 541 | |||
XM_011520625.2 | XP_011518927.1 | 5 | 1509 | |||
XM_017019212.1 | XP_016874701.1 | 4 | 1399 | |||
XM_017019213.1 | XP_016874702.1 | 4 | 1595 | |||
XM_017019214.1 | XP_016874703.1 | 5 | 1752 | |||
XM_017019215.1 | XP_016874704.1 | 2 | 2729 | |||
XM_017019216.1 | XP_016874705.1 | 4 | 858 | |||
GenBank mRNA | AK074729.1 | - | 2 | 224 | ||
BC150599.1 | - | 1 | 80 | 9056969 | ||
CR456832.1 | - | 1 | 31 | |||
DB022290.1 | - | 4 | 407 | |||
JQ308175.1 | - | 1 | 31 |
Set Membership: |
G-protein coupled receptor |