Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLNR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001507.1 | NP_001498.1 | 2 | 947 | ||
GenBank mRNA | AY603964.1 | - | 2 | 947 |
Set Membership: |
G-protein coupled receptor |