Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001508.2 | NP_001499.1 | 1 | 1083 | ||
XM_011511021.2 | XP_011509323.1 | 1 | 1685 | |||
GenBank mRNA | AF034633.1 | - | 1 | 614 | ||
AK122643.1 | - | 1 | 1083 | |||
AK314895.1 | - | 1 | 840 | |||
BC125046.1 | - | 1 | 614 | 40116106 | ||
BC125047.1 | - | 1 | 603 | |||
EU076432.1 | - | 1 | 626 | |||
FJ348258.1 | - | 1 | 614 |
Set Membership: |
G-protein coupled receptor |