Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002084.3 | NP_002075.2 | 4 | 660 | ||
GenBank mRNA | AF217787.1 | - | 4 | 447 | ||
AK310242.1 | - | 4 | 512 | |||
BC013218.1 | - | 4 | 490 | |||
BC013601.1 | - | 4 | 490 | 3882071 | ||
BC025956.1 | - | 4 | 495 | |||
BC035841.1 | - | 4 | 495 | 5748087 | ||
BC050378.1 | - | 4 | 495 | 5738272 | ||
BC066949.1 | - | 4 | 509 | |||
BI522602.1 | - | 4 | 691 | |||
D00632.1 | - | 4 | 496 | |||
X58295.1 | - | 4 | 491 |