Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030002.2 | NP_001025173.1 | 14 | 1545 | ||
NM_001242442.1 | NP_001229371.1 | 14 | 1629 | |||
NM_001242443.1 | NP_001229372.1 | 14 | 1690 | |||
NM_005310.3 | NP_005301.2 | 14 | 1700 | |||
XM_017024536.1 | XP_016880025.1 | 13 | 2819 | |||
XM_017024537.1 | XP_016880026.1 | 13 | 1579 | |||
XM_017024538.1 | XP_016880027.1 | 12 | 1779 | |||
GenBank mRNA | AB008789.1 | - | 13 | 1476 | ||
AB008790.1 | - | 12 | 1388 | |||
AK027729.1 | - | 14 | 1545 | |||
AK125544.1 | - | 14 | 1629 | |||
AK222849.1 | - | 14 | 1648 | |||
AK222870.1 | - | 14 | 1647 | |||
AK290115.1 | - | 14 | 1690 | |||
AK314368.1 | - | 14 | 1682 | |||
BC006535.2 | - | 14 | 1531 | 2824302 | ||
BT006686.1 | - | 13 | 1425 | |||
D43772.1 | - | 14 | 1644 | |||
KU178103.1 | - | 13 | 1425 | |||
KU178104.1 | - | 11 | 1462 |