Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303422.1 | NP_001290351.1 | 13 | 1467 | ||
NM_004490.2 | NP_004481.2 | 14 | 2096 | |||
XM_017003899.1 | XP_016859388.1 | 14 | 1533 | |||
GenBank mRNA | AK301961.1 | - | 13 | 1412 | ||
AK313153.1 | - | 14 | 1725 | |||
BC053559.1 | - | 14 | 1566 | 6162863 | ||
L76687.1 | - | 14 | 2095 |