Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000831.3 | NP_000822.2 | 10 | 1564 | ||
GenBank mRNA | AJ249210.1 | - | 10 | 1450 | ||
AJ299451.1 | - | 10 | 1429 | |||
U16127.1 | - | 10 | 1447 |