Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282470.2 | NP_001269399.1 | 7 | 1053 | ||
NM_001282473.2 | NP_001269402.1 | 8 | 1161 | |||
NM_014619.4 | NP_055434.2 | 8 | 1013 | |||
XM_011542784.1 | XP_011541086.1 | 7 | 871 | |||
XM_011542786.2 | XP_011541088.1 | 4 | 456 | |||
XM_017017621.1 | XP_016873110.1 | 7 | 871 | |||
XM_017017622.1 | XP_016873111.1 | 7 | 871 | |||
GenBank mRNA | AK292726.1 | - | 8 | 1000 | ||
AK314284.1 | - | 7 | 1055 | |||
BC146652.1 | - | 8 | 1111 | 40147575 | ||
BC150173.1 | - | 8 | 1111 | 40147389 | ||
S67803.1 | - | 6 | 713 |