Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NR3C1 SIRNA Assays ›
Gene Symbol:
NR3C1
Gene Name:
nuclear receptor subfamily 3 group C member 1
Gene Aliases:
GCCR, GCR, GCRST, GR, GRL
Chromosome Location:
Chr.5: 143277931 - 143435512 on Build GRCh38
UniGene ID:
Hs.122926
Species:
Human
siRNA ID
HSS104484
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NR3C1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000176.2
NP_000167.1
2
798
NM_001018074.1
NP_001018084.1
2
628
NM_001018075.1
NP_001018085.1
2
531
NM_001018076.1
NP_001018086.1
2
424
NM_001018077.1
NP_001018087.1
2
1300
NM_001020825.1
NP_001018661.1
2
798
NM_001024094.1
NP_001019265.1
2
798
NM_001204258.1
NP_001191187.1
2
798
NM_001204259.1
NP_001191188.1
2
798
NM_001204260.1
NP_001191189.1
2
798
NM_001204261.1
NP_001191190.1
2
798
NM_001204262.1
NP_001191191.1
2
798
NM_001204263.1
NP_001191192.1
2
798
NM_001204264.1
NP_001191193.1
2
798
NM_001204265.1
NP_001191194.1
2
798
XM_005268419.3
XP_005268476.1
2
528
XM_005268420.4
XP_005268477.1
2
709
XM_005268422.3
XP_005268479.1
2
590
XM_005268423.3
XP_005268480.1
2
579
XM_017009397.1
XP_016864886.1
2
424
XM_017009398.1
XP_016864887.1
2
723
GenBank mRNA
AB307716.1
-
1
306
AJ877166.1
-
2
493
AJ877167.1
-
2
368
AJ877168.1
-
2
381
AJ877169.1
-
2
455
AJ877170.1
-
3
373
AJ877171.1
-
3
449
AJ877172.1
-
3
625
AJ888004.1
-
2
417
AK223594.1
-
2
652
AK314789.1
-
2
427
AM183262.1
-
1
319
BC015610.2
-
2
746
4810424
BF966114.1
-
431
BX640610.1
-
2
489
BX648553.1
-
3
590
HQ450643.1
-
1
306
HQ450644.1
-
1
306
HQ692822.1
-
1
306
JN797823.1
-
1
306
JN797824.1
-
1
306
KU921432.1
-
1
319
U01351.1
-
2
799
X03225.1
-
2
438
X03348.1
-
2
438
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
apoptotic process
chromosome segregation
mitotic nuclear division
signal transduction
covalent chromatin modification
glucocorticoid receptor signaling pathway
glucocorticoid mediated signaling pathway
positive regulation of transcription from RNA polymerase II promoter
cell division
cellular response to steroid hormone stimulus
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
RNA binding
glucocorticoid receptor activity
steroid binding
protein binding
zinc ion binding
protein complex binding
glucocorticoid-activated RNA polymerase II transcription factor binding transcription factor activity
steroid hormone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-xzn5f:80/10.239.160.173:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline