Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000839.3 | NP_000830.2 | 5 | 2675 | ||
NM_001130063.1 | NP_001123535.1 | 3 | 1251 | |||
XM_011533635.1 | XP_011531937.1 | 4 | 4250 | |||
XM_011533636.1 | XP_011531938.1 | 4 | 3800 | |||
XM_011533637.1 | XP_011531939.1 | 4 | 2671 | |||
XM_011533638.2 | XP_011531940.1 | 5 | 2413 | |||
XM_011533639.2 | XP_011531941.1 | 5 | 2131 | |||
XM_011533640.2 | XP_011531942.1 | 5 | 2084 | |||
XM_011533641.2 | XP_011531943.1 | 5 | 2120 | |||
XM_011533643.1 | XP_011531945.1 | 5 | 1289 | |||
XM_011533644.1 | XP_011531946.1 | 4 | 1365 | |||
XM_011533645.1 | XP_011531947.1 | 4 | 1338 | |||
XM_011533646.2 | XP_011531948.1 | 4 | 1541 | |||
XM_011533647.1 | XP_011531949.1 | 5 | 1429 | |||
XM_017006271.1 | XP_016861760.1 | 5 | 3153 | |||
GenBank mRNA | AB209164.1 | - | 4 | 4916 | ||
AK289984.1 | - | 5 | 2668 | |||
AK313056.1 | - | 6 | 2810 | |||
AK315816.1 | - | 5 | 2670 | |||
AY999299.1 | - | 4 | 2472 | |||
BC041447.1 | - | 3 | 1226 | |||
BC113615.1 | - | 4 | 2459 | 8322667 | ||
BC113619.1 | - | 4 | 2459 | 8322671 | ||
BX648951.1 | - | 3 | 5866 | |||
EU432122.1 | - | 4 | 2436 | |||
L35318.1 | - | 4 | 2438 |
Set Membership: |
G-protein coupled receptor |