Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000178.3 | NP_000169.1 | 6, 7 | 712 | ||
NM_001322494.1 | NP_001309423.1 | 6, 7 | 779 | |||
NM_001322495.1 | NP_001309424.1 | 6, 7 | 1541 | |||
GenBank mRNA | AK293144.1 | - | 6, 7 | 603 | ||
AK298292.1 | - | 4, 5 | 890 | |||
AK311567.1 | - | 4, 5 | 797 | |||
AK312492.1 | - | 6, 7 | 692 | |||
BC007927.1 | - | 6, 7 | 699 | 3946317 | ||
EU668322.1 | - | 6, 7 | 699 | |||
EU794618.1 | - | 6, 7 | 699 | |||
KU178105.1 | - | 5, 6 | 613 | |||
U34683.1 | - | 6, 7 | 653 |