Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000178.3 | NP_000169.1 | 9 | 882 | ||
NM_001322494.1 | NP_001309423.1 | 9 | 949 | |||
NM_001322495.1 | NP_001309424.1 | 9 | 1711 | |||
GenBank mRNA | AB459500.1 | - | 5 | 450 | ||
AK293144.1 | - | 9 | 773 | |||
AK298292.1 | - | 7 | 1060 | |||
AK311421.1 | - | 7 | 596 | |||
AK312492.1 | - | 9 | 862 | |||
BC007927.1 | - | 9 | 869 | 3946317 | ||
EU668322.1 | - | 9 | 869 | |||
EU794618.1 | - | 9 | 869 | |||
KU178105.1 | - | 8 | 783 | |||
KU178106.1 | - | 6 | 526 | |||
U34683.1 | - | 9 | 823 |