Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001514.5 | NP_001505.1 | 2, 3 | 242 | ||
XM_011541299.1 | XP_011539601.1 | 2, 3 | 444 | |||
GenBank mRNA | AB451296.1 | - | 2, 3 | 123 | ||
AK289822.1 | - | 2, 3 | 188 | |||
BC020597.1 | - | 2, 3 | 188 | 4807599 | ||
BP379342.1 | - | 2, 3 | 242 | |||
F07886.1 | - | 2, 3 | 194 | |||
M76766.1 | - | 2, 3 | 161 | |||
S44184.1 | - | 1, 2 | 111 |