Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002095.4 | NP_002086.1 | 8 | 1064 | ||
XM_017013363.1 | XP_016868852.1 | 8 | 924 | |||
XM_017013364.1 | XP_016868853.1 | 8 | 854 | |||
GenBank mRNA | BC030572.2 | - | 8 | 1058 | 3955818 | |
S67861.1 | - | 8 | 1001 | |||
X63469.1 | - | 8 | 1053 |