Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002097.2 | NP_002088.2 | 9 | 1257 | ||
GenBank mRNA | AK057993.1 | - | 9 | 1793 | ||
AW627732.1 | - | 1 | 145 | |||
BC143711.1 | - | 8 | 1249 | |||
D32257.1 | - | 9 | 1256 | |||
U14134.1 | - | 8 | 982 | |||
U20272.1 | - | 9 | 1057 |