Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000856.5 | NP_000847.2 | 8 | 1641 | ||
NM_001130682.2 | NP_001124154.1 | 7 | 1547 | |||
NM_001130683.3 | NP_001124155.1 | 7 | 1515 | |||
NM_001130684.2 | NP_001124156.1 | 6 | 1487 | |||
NM_001130685.2 | NP_001124157.1 | 7 | 1336 | |||
NM_001130687.2 | NP_001124159.1 | 7 | 1547 | |||
NM_001256449.1 | NP_001243378.1 | 7 | 1478 | |||
XM_005262955.2 | XP_005263012.1 | 7 | 1509 | |||
XM_005262956.2 | XP_005263013.1 | 8 | 1324 | |||
XM_005262957.2 | XP_005263014.1 | 8 | 1429 | |||
XM_006714196.2 | XP_006714259.1 | 7 | 1496 | |||
XM_006714197.2 | XP_006714260.1 | 7 | 1271 | |||
XM_006714198.2 | XP_006714261.1 | 7 | 1156 | |||
XM_011531900.2 | XP_011530202.1 | 7 | 1235 | |||
GenBank mRNA | AK096588.1 | - | 7 | 1478 | ||
AK226125.1 | - | 7 | 1204 | |||
AK309950.1 | - | 7 | 1409 | |||
AK312952.1 | - | 7 | 1447 | |||
BC028384.2 | - | 6 | 1487 | 4792661 | ||
BC095395.1 | - | 7 | 1438 | |||
BX647200.1 | - | 7 | 1314 | |||
BX649180.1 | - | 7 | 1268 | |||
U58855.1 | - | 5 | 1122 | |||
X66534.1 | - | 8 | 1618 | |||
Y15723.1 | - | 8 | 1628 |