Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000856.5 | NP_000847.2 | 8 | 2090 | ||
NM_001130682.2 | NP_001124154.1 | 7 | 1996 | |||
NM_001130683.3 | NP_001124155.1 | 7 | 1964 | |||
NM_001130684.2 | NP_001124156.1 | 6 | 1936 | |||
NM_001130685.2 | NP_001124157.1 | 7 | 1785 | |||
NM_001130687.2 | NP_001124159.1 | 7 | 1996 | |||
NM_001256449.1 | NP_001243378.1 | 7 | 1927 | |||
XM_005262955.2 | XP_005263012.1 | 7 | 1958 | |||
XM_005262956.2 | XP_005263013.1 | 8 | 1773 | |||
XM_005262957.2 | XP_005263014.1 | 8 | 1878 | |||
XM_006714196.2 | XP_006714259.1 | 7 | 1945 | |||
XM_006714197.2 | XP_006714260.1 | 7 | 1720 | |||
XM_006714198.2 | XP_006714261.1 | 7 | 1605 | |||
XM_011531900.2 | XP_011530202.1 | 7 | 1684 | |||
GenBank mRNA | AK096588.1 | - | 7 | 1927 | ||
AK226125.1 | - | 7 | 1653 | |||
AK309950.1 | - | 7 | 1858 | |||
AK312952.1 | - | 7 | 1896 | |||
BC028384.2 | - | 6 | 1936 | 4792661 | ||
BC095395.1 | - | 7 | 1887 | |||
BX647200.1 | - | 7 | 1763 | |||
BX649180.1 | - | 7 | 1717 | |||
U58855.1 | - | 5 | 1571 | |||
X66534.1 | - | 8 | 2067 | |||
Y15723.1 | - | 8 | 2077 |