Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000857.3 | NP_000848.1 | 2 | 206 | ||
NM_001291951.1 | NP_001278880.1 | 2 | 206 | |||
NM_001291952.1 | NP_001278881.1 | 2 | 206 | |||
NM_001291953.1 | NP_001278882.1 | 3 | 392 | |||
NM_001291954.1 | NP_001278883.1 | 2 | 206 | |||
XM_011531901.1 | XP_011530203.1 | 2 | 120 | |||
XM_017008130.1 | XP_016863619.1 | 2 | 397 | |||
XM_017008132.1 | XP_016863621.1 | 2 | 397 | |||
GenBank mRNA | AF020340.1 | - | 2 | 89 | ||
AK296680.1 | - | 2 | 153 | |||
AK300296.1 | - | 2 | 158 | |||
AK307838.1 | - | 3 | 334 | |||
BC047620.1 | - | 2 | 120 | 5264160 | ||
DC317249.1 | - | 2 | 229 | |||
DC317683.1 | - | 2 | 213 | |||
DC342619.1 | - | 2 | 207 | |||
X66533.1 | - | 2 | 126 |