Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000857.3 | NP_000848.1 | 7 | 973 | ||
NM_001291951.1 | NP_001278880.1 | 8 | 1039 | |||
NM_001291952.1 | NP_001278881.1 | 8 | 1125 | |||
NM_001291953.1 | NP_001278882.1 | 8 | 1159 | |||
NM_001291954.1 | NP_001278883.1 | 7 | 973 | |||
NM_001291955.1 | NP_001278884.1 | 6 | 899 | |||
XM_011531901.1 | XP_011530203.1 | 7 | 887 | |||
XM_017008130.1 | XP_016863619.1 | 7 | 1164 | |||
XM_017008131.1 | XP_016863620.1 | 6 | 875 | |||
XM_017008132.1 | XP_016863621.1 | 7 | 1164 | |||
XM_017008133.1 | XP_016863622.1 | 6 | 951 | |||
GenBank mRNA | AF020340.1 | - | 7 | 856 | ||
AK296680.1 | - | 8 | 1072 | |||
AK299916.1 | - | 7 | 885 | |||
AK300296.1 | - | 8 | 991 | |||
AK307838.1 | - | 8 | 1101 | |||
AK315199.1 | - | 7 | 926 | |||
AK315944.1 | - | 6 | 880 | |||
BC047620.1 | - | 7 | 887 | 5264160 | ||
X66533.1 | - | 7 | 893 |