Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308187.1 | NP_001295116.1 | 5 | 510 | ||
NM_001308190.1 | NP_001295119.1 | 5 | 450 | |||
NM_002099.7 | NP_002090.4 | 6 | 549 | |||
XM_017008135.1 | XP_016863624.1 | 7 | 616 | |||
GenBank mRNA | AK290561.1 | - | 6 | 514 | ||
AY950614.1 | - | 6 | 398 | |||
BC005319.1 | - | 6 | 455 | 3930700 | ||
BC013328.1 | - | 6 | 436 | 4134368 | ||
DA537533.1 | - | 6 | 514 | |||
DA539682.1 | - | 6 | 550 | |||
EU338231.1 | - | 7 | 518 | |||
EU338232.1 | - | 6 | 435 | |||
EU338233.1 | - | 7 | 518 | |||
EU338234.1 | - | 5 | 342 | |||
GU347002.1 | - | 7 | 542 | |||
GU347003.1 | - | 7 | 621 | |||
GU347004.1 | - | 7 | 599 | |||
GU347005.1 | - | 2 | 121 | |||
GU347007.1 | - | 3 | 201 | |||
L07251.1 | - | 5 | 302 | |||
L07253.1 | - | 4 | 263 | |||
L31860.1 | - | 6 | 453 | |||
M36281.1 | - | 6 | 408 | |||
U00177.1 | - | 6 | 398 | |||
U00178.1 | - | 5 | 302 | |||
U00179.1 | - | 4 | 203 | |||
X08054.1 | - | 6 | 454 | |||
X51798.1 | - | 6 | 422 | |||
X57023.1 | - | 5 | 359 |