Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021957.3 | NP_068776.2 | 4 | 802 | ||
XM_006719063.3 | XP_006719126.1 | 3 | 334 | |||
XM_017019245.1 | XP_016874734.1 | 4 | 813 | |||
GenBank mRNA | AK308373.1 | - | 4 | 774 | ||
BC126310.1 | - | 4 | 572 | 8992026 | ||
BC126312.1 | - | 4 | 572 | 8992028 | ||
S70004.1 | - | 4 | 575 |