Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GZMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004131.4 | NP_004122.2 | 4 | 594 | ||
XM_011536685.2 | XP_011534987.1 | 4 | 578 | |||
GenBank mRNA | AY232654.1 | - | 4 | 559 | ||
AY232655.1 | - | 3 | 396 | |||
AY232656.1 | - | 2 | 249 | |||
AY372494.1 | - | 4 | 635 | |||
BC030195.1 | - | 4 | 550 | 5223876 | ||
CB528665.1 | - | 2 | 344 | |||
J03189.1 | - | 4 | 567 | |||
J04071.1 | - | 4 | 544 | |||
M17016.1 | - | 4 | 535 |