Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H1F0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005318.3 | NP_005309.1 | 1 | 588 | ||
GenBank mRNA | AK091372.1 | - | 2 | 378 | ||
AK312583.1 | - | 1 | 454 | |||
BC000145.1 | - | 1 | 435 | 3049108 | ||
BC029046.1 | - | 1 | 435 | 5189077 | ||
BI462123.1 | - | 1 | 589 | |||
CR456502.1 | - | 1 | 266 | |||
CR542220.1 | - | 1 | 150 |